Learn More
Eisco™ Wheatstone Bridge
Catalog No. S13004
Change view
Click to view available options
1.13 x 80 x 50mm; Two gaps
Catalog No.
S13004 1.13 x 80 x 50mm; Two gaps
1 options
Catalog No. S13004 Supplier Eisco™ Supplier No. PH1156A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Substantial bridge with brass strips mounted on hardwood base.
  • Polished hardwood base
  • Broad, heavily plated brass strips
  • Terminals with 4mm sockets provided
  • 24 SWG Constantan wire stretched along top of meter scale in mm and cm
  • Supplied with jockey
Specifications
Product Type Wheatstone Bridge
Description 24 SWG Constantan wire stretched along top of meter scale in mm and cm
Includes Brass strips mounted on hardwood base, jockey