missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ White Polypropylene Screw Caps for Glass Bottles and Vials

Catalog No. 03084113
Change view
Click to view available options
Neck Finish:
15-425
18-400
24-400
Liner Material:
Polyethylene Foam
PTFE/Silicone
No. per Case:
144/Cs.
200/Cs.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish Liner Material No. per Case
03084113 24-400 PTFE/Silicone 200/Cs.
03084119 15-425 Polyethylene Foam 144/Cs.
0298412 18-400 Polyethylene Foam 144/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03084113 Supplier DWK Life Sciences Supplier No. W224600

With clear surface for writing

  • Available with different liners (PTFE/silicone, foamed polyethylene, poly-vinyl, PTFE faced foamed polyethylene)
  • With choice of open or closed top

Caps with PTFE/silicone liner:

  • Provides heat resistance
  • Glass filled
  • Suited for EPA samples
Caps with foamed polyethylene liner:
  • Chemical resistant
  • Tight seal
Caps with poly-vinyl liner:
  • With moisture barrier
  • Suited for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Caps with PTFE faced foamed polyethylene liner:
  • Chemical resistant
  • Provides compressibility and sealing properties
  • Suited for high purity chemicals, strong acids and solvents

Specifications

Color White
Material Polypropylene
No. per Case 200/Cs.
Liner Material PTFE/Silicone
Neck Finish 24-400
Certifications/Compliance ISO 9001
For Use With (Equipment) Glass EPA vial