Learn More
DWK Life Sciences Wheaton™ White Polypropylene Screw Caps with bonded PTFE Faced Silicone Liner for E-Z Ex-Traction™ Vials
PTFE faced silicone liner ideal for storage and containment of volatile solvents, sensitive compounds or corrosive chemicals
Supplier: DWK Life Sciences W240837
Description
- Autoclavable
Chemical Identifiers
| White | |
| ISO 9001 | |
| Autoclavable |
| Polypropylene | |
| PTFE/Silicone |
Specifications
| White | |
| 500 Cs. | |
| 24-400 | |
| 20mL Vial | |
| Autoclavable |
| Polypropylene | |
| 500/Cs. | |
| ISO 9001 | |
| PTFE/Silicone |