missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ White Polypropylene Open-Top Caps w/PTFE Faced Silicone Liners for E-Z Ex-Traction™ Vials

Catalog No. 02927122
Change view
Click to view available options
Neck Finish:
13-425
15-425
22-400
24-400
No. per Case:
100 Cs.
1000 Cs.
250 Cs.
500 Cs.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish No. per Case
02927122 24-400 500 Cs.
02927115 13-425 250 Cs.
02927116 13-425 1000 Cs.
02927117 15-425 250 Cs.
02927118 15-425 1000 Cs.
02927119 22-400 100 Cs.
02927120 22-400 500 Cs.
02927121 24-400 100 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 02927122 Supplier DWK Life Sciences Supplier No. W240847

Suitable for storage and containment of volatile solvents, sensitive compounds or corrosive chemicals

  • Autoclavable

Specifications

Color White
Material Polypropylene
No. per Case 500 Cs.
No. per Pack 500/Cs.
Neck Finish 24-400
Certifications/Compliance ISO 9001
Closure Included Yes
Product Type Open Top Cap
For Use With (Equipment) Glass bottle or vial
Closure Liner PTFE/Silicone
Closure Material Polyethylene
Autoclavable Autoclavable
Show More Show Less