Learn More
DWK Life Sciences Wheaton™ White Polypropylene Caps with PTFE-Faced Foamed Polyethylene Liners
Excellent for environmental samples, pharmaceuticals and diagnostic reagents
Supplier: DWK Life Sciences 239241
Description
Excellent for environmental samples, pharmaceuticals and diagnostic reagents
- PTFE faced foamed polyethylene liner offers chemical resistance
- Polyethylene foam provides compressibility and sealing properties
- Good for high purity chemicals, strong acids, and solvents
- For usewith glass bottles or vials
- Polypropylene
- Screw cap
- White
Specifications
| White | |
| 72 Cs. | |
| 53-400 | |
| Glass bottle or vial |
| Polypropylene | |
| PTFE/Polyethylene | |
| ISO 9001 |