missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Straight Plug Style Bromobutyl rubber Stoppers for Vials and Serum Bottles
Description
- Available in choice of different materials (silicone and bromobutyl)
Not recommended for vials with a blow-back feature.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Product Type | Straight Plug Stopper |
| Material | Bromobutyl Rubber |
| Color | Gray |
| For Use With (Equipment) | Fits 1.5cm I.D., 3cm O.D. mouth of vials and serum bottles |
| Hardness | 50 Durometer |
| Diameter (Metric) Inner | 3cm |