Learn More
DWK Life Sciences Wheaton™ Straight Plug Style Silicone Stoppers for Vials and Serum Bottles
For maximum sealing of vials with straight wall glass finish
Supplier: DWK Life Sciences 224100178
Description
- Available in choice of different materials (silicone and bromobutyl)
Not recommended for vials with a blow-back feature.
Specifications
| Straight Plug Stopper | |
| Silicone | |
| 55 Durometer |
| 2 cm | |
| Fits 1.3cm I.D., 2cm O.D. mouth of vials and serum bottles | |
| Autoclavable |