missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: Straight Plug Stopper
Provide airtight covers
$961.39 - $3287.25
Specifications
| Autoclavable | Autoclavable |
|---|---|
| Product Type | Straight Plug Stopper |
| For Use With (Equipment) | Serum bottles and vials |
| Catalog Number | Mfr. No. | Color | Material | Diameter (Metric) | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Material | Diameter (Metric) | Price | Quantity | |||
|
06-451-61
|
DWK Life Sciences
224100330 |
Red | Rubber | 30 mm |
Case of 1000 Each for $1,572.52
|
|
|||
|
06-447J
|
DWK Life Sciences
W224100173 |
Gray | Butyl Rubber | 20 cm |
Case of 1000 Each for $961.39
|
|
|||
|
06-406-11D
|
DWK Life Sciences
224100072 |
Gray | Isoprene Rubber | 13 mm |
Case of 1000 Each for $3,287.25
|
|
|||
Description
- Flange-type stoppers are secured with aluminum crimp seals
- Slotted and three-prong plug stoppers are for freeze-dry applications
Specifications
| Autoclavable | |
| Straight Plug Stopper | |
| Serum bottles and vials |