missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences WHEATON™ Stoppers for Serum Bottles and Vials: 3-Leg Lyophilization Stopper
Description
Provide airtight covers
- 3-leg lyophilization plug stopper designed for freeze-drying applications
- Compatible with aluminum crimp seals for secure, airtight closure
- Gray Chlorobutyl construction with siliconized surface treatment
- Durometer rating of 46 for consistent compression and sealing performance
- Fits serum bottles and vials with 13mm inner diameter and 20mm outer diameter mouth
Specifications
Specifications
| Stopper Number | 13 |
| Diameter (Metric) Top | 20 mm |
| Diameter (Metric) Outer | 20 mm |
| Product Type | Rubber Stopper |
| Diameter (Metric) Bottom | 13 mm |
| Shape | Cylindrical |
| Sterility | Non-sterile |
| Material | Chlorobutyl Rubber |
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 2 cm |
| Show More |