Learn More
DWK Life Sciences Wheaton™ Spin Vanes for V-Vials
Magnetic stir bar for use with NextGen™ V Vials™
Supplier: DWK Life Sciences 903063
Description
- Made from PTFE
- Place stir bar in vial and use with magnetic stirrer
Chemical Identifiers
| PTFE | |
| 6 | |
| Triangle |
Specifications
| PTFE | |
| 6 | |
| Fit 2, 3, 5, and 10mL V-Vials |
| Triangle | |
| Spin Vane |