Learn More
DWK Life Sciences WHEATON™ Snap Plug Caps
Description
- Closure style: Snap plug cap for secure, reliable sealing
- Material: Polyethylene construction for chemical compatibility
- Liner material: TFE (polytetrafluoroethylene) for enhanced chemical resistance
- Available in multiple sizes and colors to suit a range of vial and tube formats
- Suitable for laboratory and clinical use
Specifications
Specifications
| Color | Natural |
| Material | LDPE |
| Sterility | Non-sterile |
| Closure Type | Snap Cap |
| Quantity | 1000/Cs. |
| Closure Size | 8 mm |
| Product Type | Plug Cap |