Learn More
DWK Life Sciences Wheaton™ Snap Plug Caps
Manufactured from Polyethylene
Supplier:  DWK Life Sciences 242804
 View more versions of this product
Catalog No. 03340111
Only null left
Description
Description
  • Closure Style: Snap Cap
  • Liner Material: TFE
Specifications
Specifications
Blue
LDPE
8 mm
Product Suggestions
Product Suggestions