Learn More
DWK Life Sciences Wheaton™ Shorty Vials™ Glass Sample Vials in Lab File™: with Caps
Come in convenient Lab File partitioned trays
Supplier:  DWK Life Sciences W224607
 View more versions of this product
Catalog No. 02912515
Only null left
Description
Description
Offering a variety of uses ranging from sample storage to freeze-drying.
  • Clear vials made from borosilicate glass
  • Fabricated from glass tubing for uniform sidewall and bottom thickness
  • Shorty Vials feature a low profile for greater stability and less dead volume
  • Supplied with attached caps to help maintain cleanliness or without caps (replacement caps can be purchased separately)
  • Vials and caps are autoclavable
  • Selected items fit M-T Vial File™—polystyrene trays with partitions that provide an easy way to inventory samples or to store empty vials
Clear Vials
  • Made from low extractable borosilicate glass to provide superior chemical resistance
  • Conform to USP Type I and ASTM E 438 Type I, Class A requirements
Cap Liners
  • 14B rubber-lined black phenolic screw cap for aqueous samples
  • PTFE/14B rubber-lined black phenolic screw cap for organic samples
Specifications
Chemical Identifiers
Clear
For uses ranging from sample storage to freeze drying
Phenolic
Autoclavable
Borosilicate Glass (Low Extractable)
Screw
Screw Cap
ASTM E438, USP Type I, Type I, Class A
Specifications
PTFE/14B Rubber
17 mm
4 mL
Screw
Phenolic
Vial Rack
ASTM E438, USP Type I, Type I, Class A
Clear
Borosilicate Glass (Low Extractable)
For uses ranging from sample storage to freeze drying
1.08 dr.
Screw Cap
Autoclavable
41 mm
Product Suggestions
Product Suggestions