Learn More
DWK Life Sciences™ Shelf Pipette Holder
Catalog No. 13709413
Change view
Click to view available options
No. per Case:
1
4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Case
13709413 1
13709414 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13709413 Supplier DWK Life Sciences™ Supplier No. W844131
Accepts two micro, macro or multichannel pipettes for Acura™ and Calibra™ systems
  • Adheres to lab bench easily and quickly
  • Noncharging
  • Durable polycarbonate construction
  • Color: Transparent Blue
Specifications
Color Blue
Material Polycarbonate
Positions 2
No. per Case 1