missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences™ Wheaton™ Serum Vial
Serum vials are converted from Type I borosilicate glass tubing.
Supplier: DWK Life Sciences™ WUBB10MLATEP
Description
- Vials conform to the hydrolytic resistance requirements outlined in USP 660 and EP 3.2.1
- Shrink-wrapped modules reduce particulate contamination
- Special design adds strength for freeze-drying applications
Chemical Identifiers
| Borosilicate Glass | |
| Non-sterile | |
| Autoclavable |
| No | |
| No | |
| Serum Vial |
Specifications
| 0.51 in. | |
| 13 mm | |
| 20 mm | |
| No | |
| 10 mL | |
| 0.34 oz. | |
| 150 Vials/Pk. | |
| Autoclavable | |
| 50 mm |
| 0.79 in. | |
| Amber | |
| Borosilicate Glass | |
| Non-sterile | |
| 1.97 in. | |
| Crimp Cap | |
| No | |
| USP Type I | |
| Serum Vial |