missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences™ Wheaton™ Serum Vial
Serum vials are converted from Type I borosilicate glass tubing.
Supplier: DWK Life Sciences™ W0EBB06RPEP
Description
- Vials conform to the hydrolytic resistance requirements outlined in USP 660 and EP 3.2.1
- Shrink-wrapped modules reduce particulate contamination
- Special design adds strength for freeze-drying applications
Chemical Identifiers
| Borosilicate Glass | |
| Non-sterile | |
| Autoclavable |
| No | |
| No | |
| Serum Vial |
Specifications
| Clear | |
| Borosilicate Glass | |
| Non-sterile | |
| 304 Vials/Pk., 100 Pk./Cs. | |
| Autoclavable | |
| Serum Vial |
| 20 mm | |
| No | |
| 10 mL | |
| No | |
| 40 mm |