Learn More
DWK Life Sciences™ Wheaton™ Serum Vial
Serum vials are converted from Type I borosilicate glass tubing.
Supplier:  DWK Life Sciences™ W0EBB10RPEP
 View more versions of this product
Catalog No. 01257624
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Vials conform to the hydrolytic resistance requirements outlined in USP 660 and EP 3.2.1
  • Shrink-wrapped modules reduce particulate contamination
  • Special design adds strength for freeze-drying applications
Specifications
Chemical Identifiers
Borosilicate Glass
Non-sterile
Autoclavable
No
No
Serum Vial
Specifications
Clear
Borosilicate Glass
Non-sterile
288 Vials/Pk., 150 Pk./Cs.
Autoclavable
Serum Vial
20 mm
No
13.5 mL
No
45 mm
Product Suggestions
Product Suggestions