missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences™ Wheaton™ Serum Vial

Catalog No. 01257611
Change view
Click to view available options
Volume (Metric):
2 mL
3 mL
4 mL
5 mL
10 mL
13.5 mL
Color:
Amber
Clear
Quantity:
10 Vials/Pk., 100 Pk./Cs.
100 Vials/Pk.
100 Vials/Pk., 10 Pk./Cs.
100 Vials/Pk., 304 Pk./Cs.
12 Vials/Pk., 196 Pk./Cs.
150 Vials/Pk.
150 Vials/Pk., 14 Pk./Cs.
150 Vials/Pk., 264 Pk./Cs.
16 Vials/Pk., 150 Pk./Cs.
169 Vials/Pk.
169 Vials/Pk., 10 Pk./Cs.
169 Vials/Pk., 304 Pk./Cs.
225 Vials/Pk.
225 Vials/Pk., 12 Pk./Cs.
225 Vials/Pk., 304 Pk./Cs.
288 Vials/Pk., 150 Pk./Cs.
304 Vials/Pk., 100 Pk./Cs.
304 Vials/Pk., 196 Pk./Cs.
30 product options available for selection
Product selection table with 30 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color Quantity
01257611 5 mL Amber 100 Vials/Pk., 10 Pk./Cs.
01671210 2 mL Amber 225 Vials/Pk.
01671211 2 mL Clear 225 Vials/Pk.
01257617 2 mL Clear 225 Vials/Pk., 12 Pk./Cs.
01257616 2 mL Clear 225 Vials/Pk., 304 Pk./Cs.
01257619 2 mL Amber 225 Vials/Pk., 12 Pk./Cs.
01257618 2 mL Amber 225 Vials/Pk., 304 Pk./Cs.
01671205 3 mL Clear 169 Vials/Pk.
01257613 3 mL Clear 169 Vials/Pk., 10 Pk./Cs.
01257612 3 mL Clear 169 Vials/Pk., 304 Pk./Cs.
01671204 3 mL Amber 169 Vials/Pk.
01257615 3 mL Amber 169 Vials/Pk., 10 Pk./Cs.
01257614 3 mL Amber 169 Vials/Pk., 304 Pk./Cs.
01257629 4 mL Clear 12 Vials/Pk., 196 Pk./Cs.
01257628 4 mL Clear 304 Vials/Pk., 196 Pk./Cs.
01671208 5 mL Clear 100 Vials/Pk.
01257609 5 mL Clear 100 Vials/Pk., 10 Pk./Cs.
01257608 5 mL Clear 100 Vials/Pk., 304 Pk./Cs.
01671206 5 mL Amber 100 Vials/Pk.
01257610 5 mL Amber 100 Vials/Pk., 304 Pk./Cs.
01671209 10 mL Clear 150 Vials/Pk.
01257621 10 mL Clear 150 Vials/Pk., 14 Pk./Cs.
01257620 10 mL Clear 150 Vials/Pk., 264 Pk./Cs.
01257627 10 mL Clear 10 Vials/Pk., 100 Pk./Cs.
01257626 10 mL Clear 304 Vials/Pk., 100 Pk./Cs.
01671207 10 mL Amber 150 Vials/Pk.
01257623 10 mL Amber 150 Vials/Pk., 14 Pk./Cs.
01257622 10 mL Amber 150 Vials/Pk., 264 Pk./Cs.
01257625 13.5 mL Clear 16 Vials/Pk., 150 Pk./Cs.
01257624 13.5 mL Clear 288 Vials/Pk., 150 Pk./Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 30 options available.
30 options
Catalog No. 01257611 Supplier DWK Life Sciences™ Supplier No. W0UBB5MLACEP
Only null left

Serum vials are converted from Type I borosilicate glass tubing.

  • Vials conform to the hydrolytic resistance requirements outlined in USP 660 and EP 3.2.1
  • Shrink-wrapped modules reduce particulate contamination
  • Special design adds strength for freeze-drying applications

Specifications

Color Amber
Diameter (Metric) Outer 20 mm
Material Borosilicate Glass
Pre-slit No
Sterility Non-sterile
Volume (Metric) 5 mL
Quantity 100 Vials/Pk., 10 Pk./Cs.
Marking Spot No
Autoclavable Autoclavable
Height (Metric) 40 mm
Product Type Serum Vial
Show More Show Less