missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Serum Vials

Catalog No. 06446D
Change view
Click to view available options
Volume (Metric):
5 mL
20 mL
Color:
Clear
Diameter (Metric) Outer:
22 mm
30.5 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color Diameter (Metric) Outer
06446D 5 mL Clear 22 mm
06446E 20 mL Clear 30.5 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06446D Supplier DWK Life Sciences Supplier No. 223685

Available as clear and amber vials

Glass can withstand the challenges of reduced pressure and extreme temperature variations. Protects against changes in pH and maintain the purity of the contents.

  • Clear vials made from low extractable borosilicate glass tubing to ensure uniform wall thickness
  • Amber vials made from borosilicate glass conforms to USP Type 1 requirements to protect light-sensitive samples
  • Withstand extreme temperature variations
  • Necks finished for aluminum crimp seals
  • Blowback feature of neck and specially designed bottom radius for lyophilization applications
  • Packaged in shrink-wrapped modules to reduce particulate contamination

Lyophilization, packaging, and sampling

Compliance

Low extractable borosilicate glass meets ASTM™ Type 1, Class A and USP Type 1 standards

Specifications

Diameter (English) Inner 0.51 in.
Diameter (Metric) Inner 13 mm
Color Clear
Diameter (Metric) Outer 22 mm
Material Borosilicate Glass
Volume (Metric) 5 mL
For Use With (Application) For lyophilization, packaging, sampling and freeze drying applications
Volume (English) 1.4 dr.
Product Line Wheaton
Volume (Metric) Total 5 mL
Closure Type Crimp Cap
Autoclavable Autoclavable
Certifications/Compliance ASTM E438, USP Type I, Type I, Class A
Height (Metric) 40 mm
Product Type Serum Vial
Show More Show Less