missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Septa For Unlined Aluminum Seals

Catalog No. 0645170
Change view
Click to view available options
Closure Size:
13 mm
20 mm
Diameter (English):
0.5 in.
0.7 in.
Color:
Gray
Red
Material:
PTFE-faced Butyl
PTFE-faced Silicone
Silicone
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size Diameter (English) Color Material
0645170 13 mm 0.5 in. Gray PTFE-faced Silicone
0645171 20 mm 0.7 in. Gray PTFE-faced Silicone
0645167 20 mm 0.7 in. Red Silicone
0644920 20 mm 0.7 in. Gray PTFE-faced Butyl
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 0645170 Supplier DWK Life Sciences Supplier No. 224172

For unlined aluminum seals

Specifications

Color Gray
Diameter (English) 0.5 in.
Material PTFE-faced Silicone
Quantity 1 Cs.
For Use With (Equipment) For unlined aluminum seals
Closure Size 13 mm
Diameter (Metric) 13 mm