Learn More
DWK Life Sciences Self-Refilling Repetitive Syringes, Microtest Manifolds
Description
- Stainless steel with PTFE stoppers, autoclavable
- Increased efficiency for inoculations, cell feedings, and washing
- Ideal for use with 24 or 96 well microplates
- Tightly fitting Luer and Luer Lock syringe nozzles
- Available in both a straight T style and an angular style with an offset hub
Specifications
Specifications
| Accuracy | ±6% |
| Autoclavable | Autoclavable |
| Material | Stainless Steel |
| Product Type | Syringe Manifold |
| Needle Point Number | 4 Channels |
| Needle Point Style | Straight |
| For Use With (Equipment) | Self refilling repetitive syringes, 24 and 96-well plates, Acura™ 865 microdispenser pipette |