missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Black Phenolic Screw Caps Without Liner

Catalog No. 14209150
Change view
Click to view available options
Thread Size (Metric):
39 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
14209150 39 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14209150 Supplier DWK Life Sciences Supplier No. 239853

Offers superior torque retention and excellent sealing characteristics

  • Poly–Seal liner is manufactured from low density polyethylene
  • Screw Cap
  • Phenolic
  • Black

It is recommended that liner be tested prior to use for leak resistant applications

Specifications

Color Black
Material Phenolic Resin
No. per Case 500 Cs.
Thread Size (Metric) 39 mm
Neck Finish 22-350
Certifications/Compliance ISO 9001
For Use With (Equipment) 651905/907 Vacules and stopper