missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Replacement Caps for Boston Round Bottles, Black Phenolic with Rubber Liners

Catalog No. 06451350
Change view
Click to view available options
Thread Size (Metric):
23 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
06451350 23 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06451350 Supplier DWK Life Sciences Supplier No. 239299
Only null left

Ideal for cell culture work.

  • 14B Rubber-lined caps feature a nontoxic white styrene-butadiene rubber liner ideal for cell culture work.

Specifications

Color Black
Material Phenolic Resin
Thread Size (Metric) 23 mm
Liner Material Rubber
Neck Finish 22-400
Product Type Replacement Cap
For Use With (Equipment) Boston Round Bottles
Autoclavable Autoclavable