missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Redi-Cap™ Phenolic Screw Caps for Redi-Pak™ Containers

Catalog No. 029847
Change view
Click to view available options
Neck Finish:
15-425
20-400
22-400
24-400
28-400
33-400
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish
029847 15-425
0298415 20-400
0298431 33-400
0298423 24-400
0298427 28-400
0298419 22-400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 029847 Supplier DWK Life Sciences Supplier No. 239250

Unique cone liner design offers superior torque retention and excellent sealing characteristics

  • Poly-Seal liner is manufactured from low density polyethylene
  • Phenolic
  • Screw cap
  • Color: Black

Specifications

Color Black
Material Phenolic Resin
No. per Case 144 Cs.
Neck Finish 15-425
Certifications/Compliance ISO 9001
For Use With (Equipment) Redi-pak™ container
Closure Liner Polyseal