missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Red Polypropylene Caps for Narrow-Mouth Containers
Description
- Leak-proof design
- Autoclavable
Specifications
Specifications
| Color | Red |
| Material | Polypropylene |
| No. per Case | 72 Cs. |
| Neck Finish | 24-410 |
| For Use With (Equipment) | Redi-pak™ leak resistant narrow mouth bottle |
| Certifications/Compliance | ISO 9001 |