missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ PVC Dropper Tips

Catalog No. 0645144
Change view
Click to view available options
Thread Size (Metric):
24 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
0645144 24 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0645144 Supplier DWK Life Sciences Supplier No. 224080

Dispenses 40 microliters per drop

  • White polypropylene overcap
  • Fits all 20mm serum finish containers

Specifications

Color White
Material PVC
No. per Case 100 Cs.
Thread Size (Metric) 24 mm
Neck Finish 24 mm
Certifications/Compliance ISO 9001
Product Type Dropper Tip
For Use With (Equipment) 20mm Serum finish container