DWK Life Sciences Wheaton™ Prescored Gold-Band™ Amber Ampules
Protect light-sensitive samples
Manufacturer: DWK Life Sciences 176792
Description
- Amber borosilicate glass
- Prescored to eliminate filing
- Gold-Band feature indicates prescoring
- Stems can be pull sealed or tip sealed
Compliance
Meet USP Type 1 and ASTM™ Type I, Class B standards
Specifications
1mL | |
Amber | |
1.05cm | |
6.7cm | |
Prescored Ampule |
USP Type 1, ASTM Type I, Class A | |
0.413 in. | |
2.63 in. | |
Borosilicate Glass |