missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Preassembled Polypropylene Caps

Catalog No. 0334475
Change view
Click to view available options
For Use With (Equipment):
12 mm x 32 mm E-Z Vials
12 mm x 32 mm Vials
15 x 45 mm Vials
Closure Size:
10-425
13-425
8-425
Material (Liner):
PTFE/Silicone
TFE/Silicone
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Closure Size Material (Liner)
0334475 12 mm x 32 mm E-Z Vials 10-425 TFE/Silicone
03340104 12 mm x 32 mm Vials 8-425 PTFE/Silicone
03344113 15 x 45 mm Vials 13-425 PTFE/Silicone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 0334475 Supplier DWK Life Sciences Supplier No. 242762
Only null left

Conveniently preassembled with septa

  • Screw Caps
  • Polypropylene
  • Black

Specifications

Material (Liner) TFE/Silicone
Color Black
Material Polypropylene
Closure Size 10-425
For Use With (Equipment) 12 mm x 32 mm E-Z Vials
Product Type Screw Cap