missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - White

Catalog No. 0343993
Change view
Click to view available options
Neck Finish:
13-425
20-410
No. per Case:
1000 Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish No. per Case
0343993 20-410 1000 Cs.
0343981 13-425 1000 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 0343993 Supplier DWK Life Sciences Supplier No. W242530A
Only null left

Screw cap made of radiation grade polypropylene

  • Packaged in case of 100 or 1000

Specifications

Color White
Material Polypropylene
No. per Case 1000 Cs.
Neck Finish 20-410
Certifications/Compliance ISO 9001
Product Type Polypropylene Cap
For Use With (Equipment) Plastic dropping bottle