missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Orange
Description
- Packaged in case of 100 or 1000
Specifications
Specifications
| Color | Orange |
| Material | Polypropylene |
| No. per Case | 1000 Cs. |
| Neck Finish | 8-425 |
| For Use With (Equipment) | Plastic dropping bottle |
| Certifications/Compliance | ISO 9001 |
| Product Type | Polypropylene Cap |