missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Natural

Catalog No. 0344024
Change view
Click to view available options
Neck Finish:
15-415
8-425
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish
0344024 8-425
0344014 15-415
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 0344024 Supplier DWK Life Sciences Supplier No. W24250101A

Screw cap made of radiation grade polypropylene

  • Packaged in case of 100 or 1000

Specifications

Color Natural
Material Polypropylene
No. per Case 1000 Cs.
Neck Finish 8-425
For Use With (Equipment) Plastic dropping bottle
Certifications/Compliance ISO 9001
Product Type Polypropylene Cap