missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Gray
Description
- Packaged in case of 100 or 1000
Specifications
Specifications
| Color | Gray |
| For Use With (Application) | Sealing Dropper Tips on Dropping Bottles, Controlled Liquid Dispensing |
| Material | Polypropylene |
| No. per Case | 100 |
| Sterility | Non-sterile |
| Neck Finish | 15-415 |
| Fit for Purpose | Certified Quality |
| For Use With (Equipment) | Wheaton (15 mm) dropper tips and Wheaton dropping bottles |
| Autoclavable | Not Autoclavable |