missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Blue

Catalog No. 06450152
Change view
Click to view available options
Neck Finish:
13-425
20-410
8-425
No. per Case:
100/Cs.
1000/Cs.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish No. per Case
06450152 8-425 100/Cs.
0343999 20-410 1000/Cs.
0344020 8-425 1000/Cs.
0343988 13-425 1000/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 06450152 Supplier DWK Life Sciences Supplier No. 242504

Screw cap made of radiation grade polypropylene

  • Packaged in case of 100 or 1000

Specifications

Color Blue
Material Polypropylene
No. per Case 100/Cs.
Neck Finish 8-425
Certifications/Compliance ISO 9001
Product Type Polypropylene Cap
For Use With (Equipment) Plastic dropping bottle