missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Polypropylene Caps for S/T Bottles

Catalog No. 12141621
Change view
Click to view available options
Color:
Black
Blue
White
Yellow
Product Type:
Cap
Open Top Cap
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type
12141621 Blue Open Top Cap
0644038 Black Open Top Cap
0644043 Black Cap
0644039 White Open Top Cap
0644044 White Cap
12141617 Yellow Open Top Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 12141621 Supplier DWK Life Sciences Supplier No. 24071604

For use with Wheaton Glass Diagnostic Bottles

  • Unlined
  • Open- and closed-top caps
  • 20-400 screw thread
  • Autoclavable

Specifications

Color Blue
For Use With (Application) Sealing Screw Neck Diagnostic Bottles; ;, Diagnostic Sample Storage, Liquid Handling
Material Polypropylene
No. per Case 300
Sterility Non-sterile
Thread Size (Metric) 20-400
Liner Material Linerless
Disposable Reusable
Fit for Purpose Certified Quality
Product Type Open Top Cap
For Use With (Equipment) Wheaton glass diagnostic bottles with thin‐flange stoppers
Autoclavable Autoclavable
Show More Show Less