missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Polypropylene Caps for S/T Bottles
Description
- Unlined
- Open- and closed-top caps
- 20-400 screw thread
- Autoclavable
Specifications
Specifications
| Color | White |
| Material | Polypropylene |
| No. per Case | 300/Cs. |
| Neck Finish | 20-400 |
| Certifications/Compliance | ISO 9001 |
| Product Type | Cap |
| For Use With (Equipment) | Screw neck diagnostic bottle |