Learn More
DWK Life Sciences Wheaton™ Polypropylene Caps for S/T Bottles
For use with Wheaton Glass Diagnostic Bottles
Supplier: DWK Life Sciences 24070601
Description
- Unlined
- Open- and closed-top caps
- 20-400 screw thread
- Autoclavable
Specifications
| Black | |
| 300/Cs. | |
| ISO 9001 | |
| Screw neck diagnostic bottle |
| Polypropylene | |
| 20-400 | |
| Cap |