Learn More
DWK Life Sciences Wheaton™ Polypropylene Caps for S/T Bottles
For use with Wheaton Glass Diagnostic Bottles
Supplier:  DWK Life Sciences 24071602
 View more versions of this product
Catalog No. 06-440-39
Only null left
Explore available promotions
Description
Description
  • Unlined
  • Open- and closed-top caps
  • 20-400 screw thread
  • Autoclavable
Specifications
Specifications
White
300/Cs.
ISO 9001
Screw neck diagnostic bottle
Polypropylene
20-400
Open Top Cap
Product Suggestions
Product Suggestions