missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Polypropylene Caps for Lab 45 Graduated Bottles

Catalog No. 0645081
Change view
Click to view available options
Color:
Blue
Red
White
Includes:
Inner Sealing Ring
PTFE/Silicone Liner
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Includes
0645081 White PTFE/Silicone Liner
0294035 Blue Inner Sealing Ring
0294034 Red Inner Sealing Ring
0645079 White Inner Sealing Ring
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 0645081 Supplier DWK Life Sciences Supplier No. 240740
Only null left

Replacement caps for Lab 45 media bottles

  • 45mm screw thread
  • Polypropylene
  • Autoclavable
  • Linerless

Specifications

Color White
Includes PTFE/Silicone Liner
Material Polypropylene
No. per Case 12 Cs.
Certifications/Compliance ISO 9001
For Use With (Equipment) Lab 45 graduated bottle
Autoclavable Autoclavable