missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Polypropylene Caps for Lab 45 Graduated Bottles

Catalog No. 0294035
Change view
Click to view available options
Color:
Blue
Red
White
Includes:
Inner Sealing Ring
PTFE/Silicone Liner
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Includes
0294035 Blue Inner Sealing Ring
0294034 Red Inner Sealing Ring
0645079 White Inner Sealing Ring
0645081 White PTFE/Silicone Liner
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 0294035 Supplier DWK Life Sciences Supplier No. 24072604
Only null left

Replacement caps for Lab 45 media bottles

  • 45mm screw thread
  • Polypropylene
  • Autoclavable
  • Linerless

Specifications

Color Blue
Includes Inner Sealing Ring
Material Polypropylene
No. per Case 12 Cs.
Certifications/Compliance ISO 9001
For Use With (Equipment) Lab 45 graduated bottle
Autoclavable Autoclavable