Learn More
DWK Life Sciences Wheaton™ Polypropylene Caps for Lab 45 Graduated Bottles
Replacement caps for Lab 45 media bottles
Supplier:  DWK Life Sciences 24072603
 View more versions of this product
Catalog No. 02-940-34
Only null left
Explore available promotions
Description
Description
  • 45mm screw thread
  • Polypropylene
  • Autoclavable
  • Linerless
Specifications
Specifications
Red
Polypropylene
ISO 9001
Autoclavable
Inner Sealing Ring
12 Cs.
Lab 45 graduated bottle
Product Suggestions
Product Suggestions