missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Polypropylene Caps For Wheaton Dropping Bottles - Green
Description
- Packaged in case of 100 or 1000
Specifications
Specifications
| Color | Green |
| Material | Polypropylene |
| No. per Case | 1000/Cs. |
| Neck Finish | 20-410 |
| Certifications/Compliance | ISO 9001 |
| Product Type | Polypropylene Cap |
| For Use With (Equipment) | Dropping bottle |