missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Polycarbonate Erlenmeyer Shake Flasks

Catalog No. 02924983
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02924983 1000 mL
02924981 250 mL
02924980 125 mL
02924982 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02924983 Supplier DWK Life Sciences Supplier No. WPFPC1000S

Ideal replacement for glass Erlenmeyer/Shake Flasks

  • Made of Lexan Polycarbonate Resin that has mechanical, optical, electrical and thermal properties
  • Supplied standard with vented, autoclavable proprietary DuoCap™ that allows gas exchange
  • Molded in graduation markings
  • Certified pyrogen, DNase and RNase free
  • Gamma irradiated to sterility assurance level (SAL) of 10-6
  • Sterile flat base flasks individually wrapped in easy to tear sterile bag
  • Flame retardant and scratch resistant resin meets USP requirements

Specifications

Certifications/Compliance Pyrogen-free, DNase-free, RNase-free, Food-grade Compliant, Manufactured in Class 10,000 Clean Room
Closure Color Blue
Closure Material Polypropylene
Color Clear
Capacity (Metric) 1000 mL
Material Polycarbonate
For Use With (Application) Suspension Cell Cultures
For Use With (Equipment) Standard Shaking Incubator Systems
No. per Pack 12/Pk.
Product Type Cell Culture Erlenmeyer Flask
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Mouth Style Threaded
Sterility Sterile
Bottom Shape Flat
Closure Type Vented DuoCap
Closure Size 38-430
Show More Show Less