Learn More
DWK Life Sciences Wheaton™ Polycarbonate Erlenmeyer Shake Flasks
Description
- Made of Lexan Polycarbonate Resin that has mechanical, optical, electrical and thermal properties
- Supplied standard with vented, autoclavable proprietary DuoCap™ that allows gas exchange
- Molded in graduation markings
- Certified pyrogen, DNase and RNase free
- Gamma irradiated to sterility assurance level (SAL) of 10-6
- Sterile flat base flasks individually wrapped in easy to tear sterile bag
- Flame retardant and scratch resistant resin meets USP requirements
Specifications
Specifications
| Closure Color | Blue |
| Closure Material | Polypropylene |
| Color | Clear |
| Capacity (Metric) | 250 mL |
| Material | Polycarbonate |
| No. per Pack | 24/Pk. |
| Closure Type | Vented DuoCap |
| Closure Size | 38-430 |