Learn More
DWK Life Sciences™ Wheaton™ Polyacetal Clamps
Color-coded clamps hold standard taper joints securely under moderate pressure
$579.61 - $633.54
| Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
|
05884C
|
DWK Life Sciences™
297746 |
Yellow |
Case of 12 Each for $579.61
|
|
|||||
|
05884D
|
DWK Life Sciences™
297749 |
Blue |
Case of 12 Each for $608.64
|
|
|||||
|
05884E
|
DWK Life Sciences™
297752 |
Dark Green |
Case of 12 Each for $633.54
|
|
|||||
Description
- Molded from polyoxymethylene (POM), an acetal resin
- Will not scratch glass and can withstand temperatures up to 150°C
- Resistant to bases and weak acids
- Not resistant to halogen gases and concentrated acids
Specifications
| Standard Taper Clamp | |
| POM Acetal |