Learn More
DWK Life Sciences WHEATON™ Peristaltic Pump Tubing: Sinkers
Description
- Sinkers designed for use with Wheaton Omnispense™ Elite and Unispense™ PRO peristaltic pumps
- Available in small and large sizes to accommodate different tubing diameters
- Borosilicate glass sinker construction
- Suitable accessory for peristaltic pump dispensing systems
Specifications
Specifications
| Diameter (Metric) Inside | 2 and 3 mm |
| For Use With (Application) | Dispensing pump |
| For Use With | Dispensing pump |
| Material | Glass |
| Size | Small |
| Tubing Size | Small |
| Type | Sinker |