missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Open Top Screw Caps with Silicone Liner

Catalog No. 03410045
Change view
Click to view available options
Neck Finish:
13-425
15-425
Color:
Black
White
No. per Case:
1000 Cs.
250 Cs.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish Color No. per Case
03410045 15-425 Black 1000 Cs.
03410041 13-425 Black 250 Cs.
03410043 13-425 Black 1000 Cs.
03410044 15-425 Black 250 Cs.
03410042 13-425 White 250 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 03410045 Supplier DWK Life Sciences Supplier No. W240851
Only null left

Open top screw caps have a bonded PTFE faced silicone liner that provides access with a syringe.

  • Can withstand autoclaving / sterilization

Specifications

Color Black
Material Phenolic Resin
No. per Case 1000 Cs.
Neck Finish 15-425
Certifications/Compliance ISO 9001
Product Type Open Top Screw Cap
For Use With (Equipment) Glass bottle or vial
Closure Liner PTFE/Silicone