Learn More
DWK Life Sciences Wheaton™ Omnispense™ Plus Peristaltic Pump Accessory Pump Heads
For use with Wheaton Omnispense Plus dispensing pump
$765.29 - $1,686.40
Specifications
For Use With Omnispense™ plus Peristaltic Pump
Holds 4
View More Specs
Products 4
Catalog Number Mfr. No. Type For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. Type For Use With (Application) Price Quantity & Availability  
13687442
View Documents Promotion Details
DWK Life Sciences
W375022
Stacking Pump Head 2.3 mm Wall Tubing
Each for $794.68
Only null left
13687443
View Documents Promotion Details
DWK Life Sciences
W375023
Pump Head 1.6 mm Wall Tubing
Each for $770.46
Only null left
13687444
View Documents Promotion Details
DWK Life Sciences
W375024
Stacking Pump Head 1.6 mm Wall Tubing
Each for $765.29
Only null left
13687445
View Documents Promotion Details
DWK Life Sciences
W375025
Cassette Pump Head Small Bore Tubing
Each for $1,686.40
Only null left
Description
Description
  • Stackable pump heads (13-687-444 and -442) allow user to add up to two additional pumpheads, for a total of three pump heads per dispensing pump
  • Cassette pump head ideal for multichannel applications
  • Removable cassettes can be preloaded with tubing elements and changed without stopping the drive
†For tubing with bore sizes from 0.13 to 2.79mm.
Specifications
Specifications
Omnispense™ plus Peristaltic Pump
4