missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Natural Polypropylene Caps for Narrow-Mouth Containers

Catalog No. 0987714A
Change view
Click to view available options
Neck Finish:
13-425
20-410
24-410
38-430
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish
0987714A 38-430
0987710A 13-425
0987711A 20-410
0987712A 24-410
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 0987714A Supplier DWK Life Sciences Supplier No. 239516

For use with Wheaton leak-resistant narrow mouth bottles

  • Leak-proof design
  • Autoclavable

Specifications

Color Natural
Material Polypropylene
No. per Case 72 Cs.
Neck Finish 38-430
Certifications/Compliance ISO 9001
For Use With (Equipment) Redi-pak™ leak resistant narrow mouth bottle