Learn More
DWK Life Sciences Modules for Acura™ Electro 956 Pipetters
Catalog No. 03410193
Change view
Click to view available options
Volume (Metric):
10 to 200 μL
2.5 to 50 μL
20 to 350 μL
Increments:
0.25 μL
1 μL
5 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Increments
03410193 20 to 350 μL 5 μL
03410191 2.5 to 50 μL 0.25 μL
03410192 10 to 200 μL 1 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03410193 Supplier DWK Life Sciences Supplier No. W870140
Interchangeable volumetric modules for Acura electro pipet line
  • Store performance data in instrument memory permanently
  • One control unit fits a choice of 27 volumetric assemblies available separately
Specifications
Model 956
Number of Channels 8
Type Interchangeable Volumetric Module
Volume (Metric) 20 to 350 μL
Finger Style Ergonomic
Increments 5 μL
Warranty 2 Years