Learn More
DWK Life Sciences Modules for Acura™ Electro 956 Pipetters
Interchangeable volumetric modules for Acura electro pipet line
Supplier: DWK Life Sciences W870138
Description
- Store performance data in instrument memory permanently
- One control unit fits a choice of 27 volumetric assemblies available separately
Specifications
| 956 | |
| Interchangeable Volumetric Module | |
| Ergonomic | |
| 2 Years |
| 8 | |
| 10 to 200 μL | |
| 1 μL |